MilliporeSigma
All Photos(1)

D3928

Sigma-Aldrich

O-(2-Oxo-1(2H)pyridyl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate

>99% (HPLC)

Synonym(s):
TPTU, O-(1,2-Dihydro-2-oxo-1-pyridyl)-N,N,N′-N′-tetramethyluronium tetrafluoroborate
Empirical Formula (Hill Notation):
C10H16BF4N3O2
CAS Number:
Molecular Weight:
297.06
MDL number:

assay

>99% (HPLC)

mp

140 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.CN(C)C(\ON1C=CC=CC1=O)=[N+](/C)C

InChI

1S/C10H16N3O2.BF4/c1-11(2)10(12(3)4)15-13-8-6-5-7-9(13)14;2-1(3,4)5/h5-8H,1-4H3;/q+1;-1

InChI key

CZQGINAUZYECAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service