MilliporeSigma
All Photos(1)

D6264

Sigma-Aldrich

Glycerol 1,3-didecanoate

~99%

Synonym(s):
1,3-Dicaprinoylglycerol, 1,3-Didecanoylglycerol, 1,3-Dicaprin
Empirical Formula (Hill Notation):
C23H44O5
Molecular Weight:
400.59
Beilstein:
1805195
MDL number:
PubChem Substance ID:

assay

~99%

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCC

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service