Documents

D6772

Sigma-Aldrich

2′-Deoxyadenosine 3′-monophosphate ammonium salt

~98%

Empirical Formula (Hill Notation):
C10H14N5O6P · xNH3
CAS Number:
Molecular Weight:
331.22 (free acid basis)
MDL number:

assay

~98%

storage temp.

−20°C

SMILES string

N.Nc1ncnc2n(cnc12)C3CC(OP(O)(O)=O)C(CO)O3

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.