MilliporeSigma
  • D8131
All Photos(1)

D8131

Sigma-Aldrich

Dansyl-γ-amino-n-butyric acid cyclohexylammonium salt

Empirical Formula (Hill Notation):
C16H20N2O4S · C6H13N
CAS Number:
Molecular Weight:
435.58
EC Number:
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

NC1CCCCC1.CN(C)c2cccc3c(cccc23)S(=O)(=O)NCCCC(O)=O

InChI

1S/C16H20N2O4S.C6H13N/c1-18(2)14-8-3-7-13-12(14)6-4-9-15(13)23(21,22)17-11-5-10-16(19)20;7-6-4-2-1-3-5-6/h3-4,6-9,17H,5,10-11H2,1-2H3,(H,19,20);6H,1-5,7H2

InChI key

AEYRCAMGXZZOLB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service