MilliporeSigma
All Photos(1)

D8408

Sigma-Aldrich

5,5-Diphenylhydantoin-d5

Synonym(s):
5-Phenyl-5-(phenyl-d5)-2,4-imidazolidinedione
Linear Formula:
C15D5H7N2O2
CAS Number:
Molecular Weight:
257.30
MDL number:

isotopic purity

98 atom % D

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2(NC(=O)NC2=O)c3ccccc3

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service