D9630

Sigma-Aldrich

Dodecanedioic acid

≥99%

Synonym(s):
Decane-1,10-dicarboxylic acid
Linear Formula:
HOOC(CH2)10COOH
CAS Number:
Molecular Weight:
230.30
Beilstein/REAXYS Number:
1782580
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

21 mmHg ( 222 °C)

assay

≥99%

bp

245 °C/10 mmHg (lit.)

mp

127-129 °C (lit.)

storage temp.

−20°C

SMILES string

OC(=O)CCCCCCCCCCC(O)=O

InChI

1S/C12H22O4/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16/h1-10H2,(H,13,14)(H,15,16)

InChI key

TVIDDXQYHWJXFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

nwg

Flash Point(F)

428 °F

Flash Point(C)

220 °C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.