All Photos(1)

E1381

Sigma-Aldrich

Erythritol bis(carbonate)

Synonym(s):
Erythritol 1,2:3,4-dicarbonate
Empirical Formula (Hill Notation):
C6H6O6
CAS Number:
Molecular Weight:
174.11
MDL number:
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[H][C@]1(COC(=O)O1)[C@@]2([H])COC(=O)O2

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service