E1635

Sigma-Aldrich

(±)-Epinephrine

≥95%

Synonym(s):
(±)-Adrenalin, 4-[1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol, (±)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, 3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol
Linear Formula:
(HO)2C6H3CH(CH2NHCH3)OH
CAS Number:
Molecular Weight:
183.20
EC Number:
MDL number:
PubChem Substance ID:

assay

≥95%

mp

197 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

CNCC(O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3

InChI key

UCTWMZQNUQWSLP-UHFFFAOYSA-N

Gene Information

rat ... Adra1a(29412), Adra2a(25083), Adrb1(24925), Adrb2(24176)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Adrenoceptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 2811 6.1 / PGII

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Kathleen G L Ma et al.
Journal of experimental zoology. Part A, Ecological and integrative physiology, 331(8), 427-442 (2019-08-07)
Doppler and B-mode ultrasonography and electrocardiography (ECG) were used to determine cardiac hemodynamics and electrical characteristics in 12°C-acclimated and metomidate-anesthetized northern pike, rainbow trout and white sturgeon (7-9 per species) at 12°C and 20°C, and at a comparable heart rate...
Aguirre A de Cubas et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(13), 3020-3030 (2015-04-01)
Pheochromocytoma and paraganglioma (PPGL) are rare neuroendocrine tumors, associated with highly variable postoperative evolution. The scarcity of reliable PPGL prognostic markers continues to complicate patient management. In this study, we explored genome-wide DNA methylation patterns in the context of PPGL...
Kyung-Won Baek et al.
Lasers in surgery and medicine, 47(5), 426-432 (2015-05-07)
Despite of the long history of medical application, laser ablation of bone tissue became successful only recently. Laser bone cutting is proven to have higher accuracy and to increase bone healing compared to conventional mechanical bone cutting. But the reason...
Catharina Göthberg et al.
Clinical implant dentistry and related research, 16(4), 487-500 (2013-01-09)
To evaluate 1-year implant survival and marginal bone loss around implants that support fixed partial dentures loaded immediately or after 3 months, and effects from abutment usage. In this 2005 to 2009 randomized, parallel-group, clinical trial, 50 partially edentulous patients...
R M Pereira et al.
Animal reproduction science, 111(1), 31-40 (2008-04-01)
A positive association between P4 concentration and initial bovine embryo survival has been reported. The objective of this study was to establish two coculture systems as a model to study the influence of progesterone on the initial bovine embryo development....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.