E2253

Sigma-Aldrich

1,N6-Ethenoadenosine 3′:5′-cyclic monophosphate sodium salt

Linear Formula:
C12H11N5O6PNa
CAS Number:
Molecular Weight:
375.21
MDL number:
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

[Na].OCC1OC(C2OP(O)(=O)OC12)n3cnc4c3ncn5ccnc45

InChI

1S/C12H12N5O6P.Na.H/c18-3-6-8-9(23-24(19,20)22-8)12(21-6)17-5-14-7-10-13-1-2-16(10)4-15-11(7)17;;/h1-2,4-6,8-9,12,18H,3H2,(H,19,20);;

InChI key

POMXMJRUTVRUAO-UHFFFAOYSA-N

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service