MilliporeSigma
All Photos(1)

E3507

Sigma-Aldrich

[D-Ala2, D-Met5]-Enkephalin acetate salt

≥97% (HPLC)

Empirical Formula (Hill Notation):
C28H37N5O7S
CAS Number:
Molecular Weight:
587.69
MDL number:
PubChem Substance ID:

assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(O)=O.CSCC[C@@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)[C@@H](C)NC(=O)[C@@H](N)Cc2ccc(O)cc2)C(O)=O

InChI

1S/C28H37N5O7S.C2H4O2/c1-17(31-26(37)21(29)14-19-8-10-20(34)11-9-19)25(36)30-16-24(35)32-23(15-18-6-4-3-5-7-18)27(38)33-22(28(39)40)12-13-41-2;1-2(3)4/h3-11,17,21-23,34H,12-16,29H2,1-2H3,(H,30,36)(H,31,37)(H,32,35)(H,33,38)(H,39,40);1H3,(H,3,4)/t17-,21+,22-,23+;/m1./s1

InChI key

PLNYJKTXNBXJNZ-UJKXDIGWSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Effects of enkephalin in analogues on prolactin release in the rat.
H Y Meltzer et al.
Life sciences, 22(21), 1931-1937 (1978-06-05)
Peptides.
Ling, N., et al. et al.
Proceedings of the Fifth American Peptide Symposium, 96-96 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service