F3511

Sigma-Aldrich

4-Fluoro-3-nitrobenzoic acid

~99%

Linear Formula:
FC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
185.11
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

~99%

mp

123-126 °C (lit.)

SMILES string

OC(=O)c1ccc(F)c(c1)[N+]([O-])=O

InChI

1S/C7H4FNO4/c8-5-2-1-4(7(10)11)3-6(5)9(12)13/h1-3H,(H,10,11)

InChI key

BOJWTAQWPVBIPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

2

J C Wijkmans et al.
Molecular diversity, 3(2), 117-120 (1997-01-01)
4-Fluoro-3-nitrobenzoic acid attached to a solid support was shown to react under mild conditions with a wide range of functionalized phenols to yield, after cleavage, the corresponding biaryl ethers in excellent purity. In a similar fashion, biaryl thioethers could be...
Wen-Bing Yeh et al.
Combinatorial chemistry & high throughput screening, 7(3), 251-255 (2004-05-12)
Liquid phasel synthesis of biheterocyclic benzimidazoles by controlled microwave irradiation was investigated. Polymer immobilized o-phenylenediamines was synthesized under microwave irradiation. The resulting PEG bound diamines was N-acylated with 4-fluoro-3-nitrobenzoic acid selectively in primary aromatic amino moiety. Nucleophilic aromatic substitution of...
D L Ladd et al.
Analytical biochemistry, 210(2), 258-261 (1993-05-01)
We have developed a new class of reagents (2) for the covalent attachment of polyethylene glycol to proteins. These reagents (2) are the monomethoxypolyethylene glycol esters of 4-fluoro-3-nitrobenzoic acid. The reaction of 2 with lysine epsilon-amino groups produces a chromophore...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.