MilliporeSigma
All Photos(1)

F5017

Sigma-Aldrich

Foroxymithine microbial

≥85% (HPLC)

Empirical Formula (Hill Notation):
C22H37N7O11
CAS Number:
Molecular Weight:
575.57
MDL number:
PubChem Substance ID:

assay

≥85% (HPLC)

SMILES string

CC(=O)NC(CCCN(O)C=O)C(=O)NC(CO)C(=O)N(O)CCCC1NC(=O)C(CCCN(O)C=O)NC1=O

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service