All Photos(1)

G6768

Sigma-Aldrich

β-D-Glucopyranosylphenyl isothiocyanate

Synonym(s):
Isothiocyanatophenyl β-D-glucopyranoside
Empirical Formula (Hill Notation):
C13H15NO6S
CAS Number:
Molecular Weight:
313.33
MDL number:
PubChem Substance ID:

biological source

synthetic

assay

≥98% (TLC)

form

powder

application(s)

thin layer chromatography (TLC): suitable

suitability

suitable for preparation of neoglycoproteins

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(cc2)N=C=S)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C13H15NO6S/c15-5-9-10(16)11(17)12(18)13(20-9)19-8-3-1-7(2-4-8)14-6-21/h1-4,9-13,15-18H,5H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

RWANFUZQWINQBY-UJPOAAIJSA-N

Certificate of Analysis

Certificate of Origin

M Monsigny et al.
Biology of the cell, 51(2), 187-196 (1984-01-01)
The presence and the sugar specificity of membrane lectins on the cell surface of mouse L1210 leukemia cells were demonstrated by using various neoglycoproteins (glycosylated serum albumin) substituted with fluorescein or methotrexate. Neoglycoproteins were prepared by reaction of glycosidophenylisothiocyanates with...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service