H4411
Sigma-Aldrich
HybChecker™
Pricing and availability is not currently available.
storage temp.
−20°C
General description
An optimized hybridization solution containing fluorescent labeled random oligonucleotides to be used for quality control evaluation of printed microarray slides. Designed to verify that the nucleic acid is present on the slide′s surface and is available for hybridization. Allows for assessment of spot morphology and detection of any spotting irregularities.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.