MilliporeSigma
  • H9884
All Photos(1)

H9884

Sigma-Aldrich

Hypothalamus acetone powder bovine

biological source

bovine

form

powder

storage temp.

2-8°C

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service