I1386

Sigma-Aldrich

Anti-Human IgD (δ-chain specific) antibody produced in goat

whole antiserum

MDL number:
Pricing and availability is not currently available.

biological source

goat

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

application(s)

immunoelectrophoresis: 1:8

conjugate

unconjugated

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IGHD(3495)

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn

Risk Statement

22

RIDADR

NONH for all modes of transport

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.