Documents

I4502

Sigma-Aldrich

2′,3′-O-Isopropylideneinosine

≥98% (HPLC)

Empirical Formula (Hill Notation):
C13H16N4O5
CAS Number:
Molecular Weight:
308.29
EC Number:
MDL number:

assay

≥98% (HPLC)

SMILES string

CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)n3cnc4C(=O)NC=Nc34

Looking for similar products? Visit Product Comparison Guide

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.