Documents

I8404

Sigma-Aldrich

Isodesmosine chloride

from bovine neck ligament, 99.5%±0.2%

Empirical Formula (Hill Notation):
C24H40ClN5O8
CAS Number:
Molecular Weight:
562.06
Pricing and availability is not currently available.

biological source

bovine neck ligament

assay

99.5%±0.2%

storage temp.

2-8°C

SMILES string

[Cl-].N[C@@H](CCCC[n+]1cc(CC[C@H](N)C(O)=O)cc(CC[C@H](N)C(O)=O)c1CCC[C@H](N)C(O)=O)C(O)=O

Caution

Extremely hygroscopic, should be stored over silica gel

Preparation Note

Each vial contains 1μmole of purified isodesmosine isolated by solvent extraction and chromatography by method of Starcher et al, and calibrated by UV and amino acid analysis

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
B C Starcher et al.
Preparative biochemistry, 5(5-6), 455-460 (1975-01-01)
A rapid large scale procedure was devised for the purification of desmosine and isodesmosine from ligamentum nuchae elastin. The method makes use of the hydrophilic nature of the desmosines which preferentially absorbs to cellulose fibers in mixtures of organic solvents....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.