MilliporeSigma
All Photos(1)

M6904

Sigma-Aldrich

Mercury(II) iodide red

ACS reagent, ≥99.0% dry basis

Synonym(s):
Mercuric iodide red
Empirical Formula (Hill Notation):
HgI2
CAS Number:
Molecular Weight:
454.40
EC Number:
MDL number:

grade

ACS reagent

assay

≥99.0% dry basis

solubility

potassium iodide solution: passes test

cation traces

Hg+: ≤0.1%
soluble mercury salts (as Hg): ≤0.05%

SMILES string

I[Hg]I

InChI

1S/Hg.2HI/h;2*1H/q+2;;/p-2

InChI key

YFDLHELOZYVNJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service