All Photos(1)

M8529

Sigma-Aldrich

N-(Methyl)mercaptoacetamide

Synonym(s):
N-(Methyl)thioglycolamide
Linear Formula:
HSCH2CONHCH3
CAS Number:
Molecular Weight:
105.16
Beilstein:
1740652
MDL number:
PubChem Substance ID:

density

1.19 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CNC(=O)CS

InChI

1S/C3H7NOS/c1-4-3(5)2-6/h6H,2H2,1H3,(H,4,5)

InChI key

NSJNRJYQQPRCLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Mild reducing agent for methionine sulfoxide.

Certificate of Analysis

Certificate of Origin

Lundblad, R.I. and Noyes, C.M., ed.
Chemical Reagents for Protein Modification, 1, 99-99 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service