N127

Sigma-Aldrich

S-(4-Nitrobenzyl)-6-thioinosine

Synonym(s):
NBTI, NBMPR, 6-[(4-Nitrobenzyl)thio]-9-β-D-ribofuranosylpurine
Empirical Formula (Hill Notation):
C17H17N5O6S
CAS Number:
Molecular Weight:
419.41
EC Number:
MDL number:

mp

187-190 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(SCc4ccc(cc4)[N+]([O-])=O)ncnc23

InChI

1S/C17H17N5O6S/c23-5-11-13(24)14(25)17(28-11)21-8-20-12-15(21)18-7-19-16(12)29-6-9-1-3-10(4-2-9)22(26)27/h1-4,7-8,11,13-14,17,23-25H,5-6H2/t11-,13-,14-,17-/m1/s1

InChI key

DYCJFJRCWPVDHY-LSCFUAHRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of equilibrative nucleoside transporters (ENTs), particularly adenosine transporters, in central nervous system and vascular smooth muscle.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.