All Photos(1)

N3758

Sigma-Aldrich

N-(3-Nitrobenzyloxymethyl)pyridinium chloride

Linear Formula:
C13H13N2O3Cl
CAS Number:
Molecular Weight:
280.71
EC Number:
MDL number:
PubChem Substance ID:

assay

98-99% (TLC)

storage temp.

2-8°C

SMILES string

[Cl-].[O-][N+](=O)c1cccc(COC[n+]2ccccc2)c1

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service