MilliporeSigma
All Photos(2)

N4765

Sigma-Aldrich

N-(3-Nitro-2-pyridinesulfenyl)-L-leucine

Empirical Formula (Hill Notation):
C11H15N3O4S
CAS Number:
Molecular Weight:
285.32
MDL number:
PubChem Substance ID:
NACRES:
NA.26

form

solid

Quality Level

storage temp.

−20°C

SMILES string

CC(C)CC(NSc1ncccc1N(=O)=O)C(O)=O

InChI

1S/C11H15N3O4S/c1-7(2)6-8(11(15)16)13-19-10-9(14(17)18)4-3-5-12-10/h3-5,7-8,13H,6H2,1-2H3,(H,15,16)

InChI key

QCVUBDSGNDCAAO-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service