O0506

Sigma-Aldrich

Orn-β-Ala dihydrochloride

Empirical Formula (Hill Notation):
C8H17N3O3 · 2HCl
CAS Number:
Molecular Weight:
276.16
MDL number:

storage temp.

−20°C

SMILES string

NCCCC(N)C(=O)NCCC(O)=O

Other Notes

"Salty" peptide

Certificate of Analysis

Certificate of Origin

Okai, H. and Tamuya, M.
Chemical and Engineering News, 68(2), 26-26 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service