P202

Sigma-Aldrich

Pituitary adenylate cyclase activating polypeptide-27 ovine

Synonym(s):
PACAP-27
Empirical Formula (Hill Notation):
C142H224N40O39S
CAS Number:
Molecular Weight:
3147.61
MDL number:

SMILES string

CCC(C)C(NC(=O)CNC(=O)C(CC(O)=O)NC(=O)C(CO)NC(=O)C(N)Cc1cnc[nH]1)C(=O)NC(Cc2ccccc2)C(=O)NC(C(C)O)C(=O)NC(CC(O)=O)C(=O)NC(CO)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(CO)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4ccc(O)cc4)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCCN)C(=O)NC(CCC(N)=O)C(=O)NC(CCSC)C(=O)NC(C)C(=O)NC(C(C)C)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(Cc5ccc(O)cc5)C(=O)NC(CC(C)C)C(=O)NC(C)C(=O)NC(C)C(=O)NC(C(C)C)C(=O)NC(CC(C)C)C(N)=O

InChI

1S/C142H224N40O39S/c1-16-75(10)113(179-108(191)66-156-120(201)102(63-109(192)193)173-135(216)104(67-183)176-119(200)88(146)62-84-65-153-70-157-84)139(220)174-101(58-80-28-18-17-19-29-80)133(214)182-114(79(14)186)140(221)175-103(64-110(194)195)132(213)178-106(69-185)136(217)172-100(61-83-39-45-87(189)46-40-83)131(212)177-105(68-184)134(215)164-93(34-27-54-155-142(151)152)126(207)170-98(59-81-35-41-85(187)42-36-81)129(210)163-92(33-26-53-154-141(149)150)124(205)161-89(30-20-23-50-143)122(203)165-94(47-48-107(147)190)127(208)166-95(49-55-222-15)121(202)159-78(13)118(199)180-111(73(6)7)137(218)167-91(32-22-25-52-145)123(204)162-90(31-21-24-51-144)125(206)171-99(60-82-37-43-86(188)44-38-82)130(211)169-97(57-72(4)5)128(209)160-76(11)116(197)158-77(12)117(198)181-112(74(8)9)138(219)168-96(115(148)196)56-71(2)3/h17-19,28-29,35-46,65,70-79,88-106,111-114,183-189H,16,20-27,30-34,47-64,66-69,143-146H2,1-15H3,(H2,147,190)(H2,148,196)(H,153,157)(H,156,201)(H,158,197)(H,159,202)(H,160,209)(H,161,205)(H,162,204)(H,163,210)(H,164,215)(H,165,203)(H,166,208)(H,167,218)(H,168,219)(H,169,211)(H,170,207)(H,171,206)(H,172,217)(H,173,216)(H,174,220)(H,175,221)(H,176,200)(H,177,212)(H,178,213)(H,179,191)(H,180,199)(H,181,198)(H,182,214)(H,192,193)(H,194,195)(H4,149,150,154)(H4,151,152,155)

InChI key

RZGBUJXSKLDAFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service