MilliporeSigma
All Photos(1)

P4898

Sigma-Aldrich

Phe-Met-Arg-Phe amide

≥95% (HPLC)

Synonym(s):
FMRF amide, Molluscan cardioexcitatory neuropeptide
Empirical Formula (Hill Notation):
C29H42N8O4S
CAS Number:
Molecular Weight:
598.76
MDL number:
PubChem Substance ID:

assay

≥95% (HPLC)

form

powder

composition

Peptide content, ~65%

technique(s)

mass spectrometry (MS): suitable

color

white

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(N)Cc1ccccc1)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc2ccccc2)C(N)=O

InChI

1S/C29H42N8O4S/c1-42-16-14-23(35-26(39)21(30)17-19-9-4-2-5-10-19)28(41)36-22(13-8-15-34-29(32)33)27(40)37-24(25(31)38)18-20-11-6-3-7-12-20/h2-7,9-12,21-24H,8,13-18,30H2,1H3,(H2,31,38)(H,35,39)(H,36,41)(H,37,40)(H4,32,33,34)

InChI key

WCSPDMCSKYUFBX-UHFFFAOYSA-N

Related Categories

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

SDS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service