MilliporeSigma
All Photos(1)

P7285

Sigma-Aldrich

1,2-Dipentadecanoyl-sn-glycero-3-phosphocholine

~99%

Synonym(s):
L-α-Phosphatidylcholine, dipentadecanoyl
Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
MDL number:
PubChem Substance ID:

assay

~99%

functional group

phospholipid

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCC)=O)=O

InChI

1S/C38H77NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-37(40)44-34-36(35-46-48(42,43)45-33-32-39(3,4)5)47-38(41)31-29-27-25-23-21-19-17-15-13-11-9-7-2/h36H,6-35H2,1-5H3,(H,42,43)

InChI key

IEYQCOTZLONQTQ-UHFFFAOYSA-N

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

J J Dominguez Pardo et al.
Chemistry and physics of lipids, 208, 58-64 (2017-09-20)
Styrene-maleic acid copolymers (SMA) have been gaining interest in the field of membrane research due to their ability to solubilize membranes into nanodics. The SMA molecules act as an amphipathic belt that surrounds the nanodiscs, whereby the hydrophobic styrene moieties
Andrea Palyzová et al.
Journal of chromatography. A, 1635, 461708-461708 (2020-11-24)
A combination of two chromatographic and two enzymatic methods was used for the analysis of molecular species of lipids from Gram-positive bacteria of the genus Kocuria. Gram-positive bacteria contain a majority of branched fatty acids (FAs), especially iso- and/or anteiso-FAs.
J Mou et al.
Biochemistry, 35(10), 3222-3226 (1996-03-12)
Using an atomic force microscope, supported bilayers of saturated phosphatidylcholine (in the gel state) containing various amounts of gramicidin A (gA) were imaged in aqueous solutions and at room temperature. gA clusters were directly observed for the first time under
Qing Wang et al.
Biophysical journal, 115(4), 664-678 (2018-08-08)
Using Förster resonance energy transfer, raft/liquid-ordered-domain formation was assessed in asymmetric vesicles containing outer leaflets composed of high-Tm (melting temperature) saturated phosphatidylcholines (diC18:0PC, diC16:0PC, diC15:0PC, or diC14:0PC), low-Tm unsaturated dioleoylphosphatidylcholine (DOPC) and cholesterol, and inner leaflets composed of lipids that
Andrea de la Garza Puentes et al.
International journal of food sciences and nutrition, 66(8), 936-942 (2015-10-31)
Plasma is the most commonly employed matrix for analyzing fatty acids (FAs), but its extraction is not well accepted in the infant population. The objectives of this study were to evaluate cheek cells and capillary blood as alternatives to plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service