MilliporeSigma
All Photos(1)

P8409

Sigma-Aldrich

1-Pyrenedecanoic acid

Synonym(s):
10-(1-Pyrenyl)decanoic acid
Empirical Formula (Hill Notation):
C26H28O2
CAS Number:
Molecular Weight:
372.50
Beilstein:
2007670
MDL number:

storage temp.

−20°C

SMILES string

OC(=O)CCCCCCCCCc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C26H28O2/c27-24(28)12-7-5-3-1-2-4-6-9-19-13-14-22-16-15-20-10-8-11-21-17-18-23(19)26(22)25(20)21/h8,10-11,13-18H,1-7,9,12H2,(H,27,28)

InChI key

PIMKEUIWMFJVNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service