P8545

Sigma-Aldrich

Pipenzolate bromide

powder

Synonym(s):
Benzilic acid 1-ethylpiperidin-3-yl ester methobromide
Linear Formula:
C22H28NO3Br
CAS Number:
Molecular Weight:
434.37
EC Number:
MDL number:
PubChem Substance ID:

form

powder

color

white

SMILES string

Br.CC[N]1(C)CCCC(C1)OC(=O)C(O)(c2ccccc2)c3ccccc3

InChI

1S/C22H28NO3.BrH/c1-3-23(2)16-10-15-20(17-23)26-21(24)22(25,18-11-6-4-7-12-18)19-13-8-5-9-14-19;/h4-9,11-14,20,25H,3,10,15-17H2,1-2H3;1H

InChI key

PRMDDJWOUVPRIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Pharmacologic effect of somatostatin on bile formation in the dog. Enhanced ductular reabsorption as the major mechanism of anticholeresis.
E René et al.
Gastroenterology, 84(1), 120-129 (1983-01-01)
Return to Pakistan of pipenzolate plus phenobarbitone.
K I Tahir
Lancet (London, England), 339(8791), 498-498 (1992-02-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.