Documents

S1821

Sigma-Aldrich

SC-68376

≥98% (HPLC), solid

Synonym(s):
2-Methyl-4-phenyl-5-(4-pyridyl)oxazole
Empirical Formula (Hill Notation):
C15H12N2O
CAS Number:
Molecular Weight:
236.27
MDL number:
PubChem Substance ID:

assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: 18 mg/mL

storage temp.

2-8°C

SMILES string

Cc1nc(-c2ccccc2)c(o1)-c3ccncc3

Biochem/physiol Actions

SC-68376 is a potent, selective inhibitor of p38 MAP kinase activity in a dose-dependent manner (IC50 = 2-5 μM).

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.