All Photos(1)

S8258

Sigma-Aldrich

Ser-Tyr

Empirical Formula (Hill Notation):
C12H16N2O5
CAS Number:
Molecular Weight:
268.27
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

O=C(N[C@@H](CC1=CC=C(C=C1)O)C(O)=O)[C@H](CO)N

InChI

1S/C12H16N2O5/c13-9(6-15)11(17)14-10(12(18)19)5-7-1-3-8(16)4-2-7/h1-4,9-10,15-16H,5-6,13H2,(H,14,17)(H,18,19)

InChI key

MALNXHYEPCSPPU-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service