SML0019

Sigma-Aldrich

VU0357121

≥98% (HPLC)

Synonym(s):
VU0357121, 4-Butoxy-N-(2,4-difluorophenyl)-benzamide
Empirical Formula (Hill Notation):
C17H17F2NO2
CAS Number:
Molecular Weight:
305.32
MDL number:
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

assay

≥98% (HPLC)

form

powder

solubility

DMSO: ≥18 mg/mL

storage temp.

2-8°C

SMILES string

CCCCOc1ccc(cc1)C(=O)Nc2ccc(F)cc2F

InChI

1S/C17H17F2NO2/c1-2-3-10-22-14-7-4-12(5-8-14)17(21)20-16-9-6-13(18)11-15(16)19/h4-9,11H,2-3,10H2,1H3,(H,20,21)

InChI key

AHCYOTLTLQTPSU-UHFFFAOYSA-N

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

VU0357121 is a positive allosteric modulator (PAM) of the metabotropic glutamate receptor 5 (mGlu5) with EC50 = 33 nM, 92% Glumax. Its site of action is distinct from the classical mGlu5 negative allosteric modulator (NAM) MPEP allosteric site or the site of a recetly discovered PAM, CPPHA. However, VU0357121shares a functional interaction with the MPEP site. VU0357121 is highly selective for activation of mGlu5 and is inactive or very weakly antagonizing at other mGlu receptor subtypes.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

UN 3077 9 / PGIII

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.