T0644

Sigma-Aldrich

5,10,15,20-Tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate)

Synonym(s):
TMPyP, meso-Tetra(N-methyl-4-pyridyl)porphine tetratosylate salt
Empirical Formula (Hill Notation):
C72H66N8O12S4
CAS Number:
Molecular Weight:
1363.60
Beilstein/REAXYS Number:
6049195
MDL number:

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.Cc2ccc(cc2)S([O-])(=O)=O.Cc3ccc(cc3)S([O-])(=O)=O.Cc4ccc(cc4)S([O-])(=O)=O.C[n+]5ccc(cc5)-c6c7ccc(n7)c(-c8cc[n+](C)cc8)c9ccc([nH]9)c(-c%10cc[n+](C)cc%10)c%11ccc(n%11)c(-c%12cc[n+](C)cc%12)c%13ccc6[nH]%13

InChI

1S/C44H37N8.4C7H8O3S/c1-49-21-13-29(14-22-49)41-33-5-7-35(45-33)42(30-15-23-50(2)24-16-30)37-9-11-39(47-37)44(32-19-27-52(4)28-20-32)40-12-10-38(48-40)43(36-8-6-34(41)46-36)31-17-25-51(3)26-18-31;4*1-6-2-4-7(5-3-6)11(8,9)10/h5-28H,1-4H3,(H,45,46,47,48);4*2-5H,1H3,(H,8,9,10)/q+3;;;;/p-3/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;;;

InChI key

AKZFRMNXBLFDNN-GVHKZQBISA-K

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Transition-metal complexes of this compound are site-specific DNA cleavage reagents.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.