MilliporeSigma
All Photos(1)

T0661

Sigma-Aldrich

Tyramine-7-14C hydrochloride

~95% (Radiochemical Purity), aqueous ethanol solution

Linear Formula:
HOC6H414CH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
173.64
MDL number:
PubChem Substance ID:

assay

~95% (Radiochemical Purity)

form

aqueous ethanol solution

extent of labeling

1-25 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

Cl.NC[14CH2]c1ccc(O)cc1

Physical form

Aqueous solution containing 2% ethanol in sealed amber ampule

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service