T5388

Sigma-Aldrich

DL-Tryptophanol oxalate salt

Synonym(s):
β-Aminoindole-3-propanol
Empirical Formula (Hill Notation):
C11H14N2O · C2H2O4
CAS Number:
Molecular Weight:
280.28
MDL number:
PubChem Substance ID:

storage temp.

−20°C

SMILES string

OC(=O)C(O)=O.NC(CO)Cc1c[nH]c2ccccc12

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

E L Paley et al.
Experimental cell research, 195(1), 66-78 (1991-07-01)
Bovine kidney cell lines resistant to tryptamine and tryptophanol (tryptophan analogs) were selected. The content of tryptophanyl-tRNA synthetase (WRS, EC 6.1.1.2) was assayed by measuring the binding of monospecific polyclonal antibodies to the 35S-labeled enzyme in detergent-soluble and -insoluble forms...
Ashley A Reinke et al.
Chembiochem : a European journal of chemical biology, 11(13), 1889-1895 (2010-08-03)
In Alzheimer's disease (AD) and other neurodegenerative disorders, proteins accumulate into ordered aggregates, called amyloids. Recent evidence suggests that these structures include both large, insoluble fibrils and smaller, prefibrillar structures, such as dimers, oligomers, and protofibrils. Recently, focus has shifted...
G M Beck et al.
Chirality, 8(7), 503-510 (1996-01-01)
Lambda-carrageenan, a linear high molecular weight sulfated polysaccharide, has been employed as a chiral selector in capillary electrophoresis for the separation of enantiomers of weakly basic pharmaceutical compounds. The racemic compounds that were enantioresolved included propranolol, pindolol, tryptophanol, laudanosine and...
Mercedes Amat et al.
The Journal of organic chemistry, 74(3), 1205-1211 (2008-12-17)
The enantioselective construction of the 3-ethylindolo[2,3-a]quinolizidine moiety present in numerous indole alkaloids is reported, the key steps being a stereoselective cyclocondensation of (S)-tryptophanol with an appropriate racemic delta-oxoester and a regio- and stereoselective cyclization of the resulting oxazolopiperidones on the...
Nicholas A Barnes et al.
Journal of immunology (Baltimore, Md. : 1950), 183(9), 5768-5777 (2009-10-16)
Catabolism of tryptophan by IDO1 plays an important role in the control of immune responses. Activation of the eukaryotic initiation factor 2alpha (eIF2alpha) kinase general control nonderepressible-2 (GCN2) following tryptophan depletion is a major pathway mediating this effect. However, immunomodulatory...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.