MilliporeSigma
  • T5908
All Photos(1)

T5908

Sigma-Aldrich

Lithium p-toluenesulfinate

≥98% (titration)

Synonym(s):
p-Toluenesulfinic acid lithium salt
Linear Formula:
CH3C6H4SO2Li
CAS Number:
Molecular Weight:
162.14
Beilstein:
4163977
EC Number:
MDL number:

assay

≥98% (titration)

SMILES string

[Li+].Cc1ccc(cc1)S([O-])=O

InChI

1S/C7H8O2S.Li/c1-6-2-4-7(5-3-6)10(8)9;/h2-5H,1H3,(H,8,9);/q;+1/p-1

InChI key

MSUZXYWQEDRGFN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service