MilliporeSigma
All Photos(1)

T8881

Sigma-Aldrich

β-(1,2,4-Triazol-3-yl)-DL-alanine

Synonym(s):
DL-1,2,4-Triazole-3-alanine, (±)-2-Amino-3-(1,2,4-triazol-3-yl)propionic acid
Empirical Formula (Hill Notation):
C5H8N4O2
CAS Number:
Molecular Weight:
156.14
EC Number:
MDL number:

storage temp.

−20°C

SMILES string

NC(Cc1nc[nH]n1)C(O)=O

InChI

1S/C5H8N4O2/c6-3(5(10)11)1-4-7-2-8-9-4/h2-3H,1,6H2,(H,10,11)(H,7,8,9)

InChI key

CAPORZWUTKSILW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service