U7500

Sigma-Aldrich

4-Imidazoleacrylic acid

≥99%

Synonym(s):
3-(4-Imidazolyl)acrylic acid, Urocanic acid
Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
Beilstein/REAXYS Number:
81405
EC Number:
MDL number:

assay

≥99%

mp

226-228 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\c1c[nH]cn1

InChI

1S/C6H6N2O2/c9-6(10)2-1-5-3-7-4-8-5/h1-4H,(H,7,8)(H,9,10)/b2-1+

InChI key

LOIYMIARKYCTBW-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis

Certificate of Origin

Silke Hack et al.
European journal of medicinal chemistry, 46(5), 1483-1498 (2011-03-01)
A new series of potential GABA uptake inhibitors starting from of 1H-imidazol-4-ylacetic acid with the carboxylic acid side chain originating from different positions and varying in length have been synthesized and tested for the inhibitory potency at the four GABA...
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.