Documents
Z365904
Hybri-slips
L × W × thickness 22 mm × 22 mm × 0.25 mm
Pricing and availability is not currently available.
L × W × thickness
22 mm × 22 mm × 0.25 mm
General description
  • Constant probe concentration - Unlike glass cover slips, Hybri-slips do not adsorb probes.
  • Ready-to-use - Hybri-slips do not require pre-treatment prior to use.
  • Avoid RNA degradation - Hybri-slips are supplied RNAse-free and are protected from contamination by clean-release liners.
  • Durable - Hybri-slips remain flat even at elevated temperatures.
Application
Hybri-slips are hydrophobic cover slips for in situ hybridization, in situ PCR, and hybridization to genomic arrays on glass slides.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.