Documents
Z378623
Double-ended pestle for microcentrifuge tubes
L 152 mm (6 in.), pkg of 1 ea, flat bottom tip
NACRES:
NB.22
Pricing and availability is not currently available.
material
flat bottom tip
packaging
pkg of 1 ea
mfr. no.
Bel-Art
L
152 mm (6 in.)
General description
The 3.2 mm stainless steel shaft carries a nylon pestle at each end. One fits standard 0.5 mL microcentrifuge tubes, the other 1.5 mL tubes. Overall length is 152 mm. May be hand-held or mounted in a motor-driven chuck.
Certificate of Analysis
Certificate of Origin
Contact Technical Service
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.