Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0479

Sigma-Aldrich

Adrafinil

≥98% (HPLC), solid

Synonym(s):

2-[(Diphenylmethyl)sulfinyl]acetohydroxamic acid, CRL 40028, Olmifon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO3S
CAS Number:
Molecular Weight:
289.35
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white

solubility

DMSO: 35 mg/mL

SMILES string

ONC(=O)CS(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C15H15NO3S/c17-14(16-18)11-20(19)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15,18H,11H2,(H,16,17)

InChI key

CGNMLOKEMNBUAI-UHFFFAOYSA-N

Biochem/physiol Actions

Agent promoting wakefulness; analog of Modafinil.

Caution

Hygroscopic

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hascoet et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 5(4), 481-489 (1995-12-01)
The binding and locomotor profile of a new sigma ligand, S14905, (isobutyl-N-(1-indan-2yl-piperid-4-yl)N-methyl carbamate, furamate) was studied. The binding data revealed that S14905 has a high affinity for sigma receptors and very low affinity for both dopamine D1 and D2 receptors.
R Nageswara Rao et al.
Biomedical chromatography : BMC, 23(8), 811-816 (2009-04-09)
A simple and rapid normal-phase HPLC method for enantiospecific separation of a psychostimulant, adrafinil (ADL), and its metabolite modafinil (MDL) in rat serum and urine was developed. The separation was accomplished on a normal-phase polysaccharide stationary phase Chiralcel OJ-H using
F Lechin et al.
Journal of medicine, 31(5-6), 333-361 (2001-08-18)
Neurochemical, neuroautonomic and neuropharmacological assessments carried out on all our myasthenia gravis (MG) patients showed that they presented a neural sympathetic deficit plus excessive adrenal-sympathetic activity. These abnormalities were registered during the basal (supine-resting) state, as well as after several
C Rozé et al.
Archives internationales de pharmacodynamie et de therapie, 265(1), 119-127 (1983-09-01)
The drug CRL 40 028 increases spontaneous motility through an action on central adrenergic receptors. The effects of this drug have been tested in rats on the external pancreatic secretion induced by secretin, CCK, acetylcholine, vagal electrical stimulation or 2
C L Milhaud et al.
Journal de pharmacologie, 16(4), 372-380 (1985-10-01)
The nocturnal activity of a primate was used as an evaluation criterium for a stimulating substance: adrafinil (CRL 40028). Ten rhesus monkeys were placed in a controlled environment and their activity was measured, in relative time, using an ultra-sound system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service