A7955

Sigma-Aldrich

Aluminum acetylacetonate

99% (titration)

Synonym(s):
Aluminum 2,4-pentanedionate, Al(acac)3
Linear Formula:
Al(C5H7O2)3
CAS Number:
Molecular Weight:
324.31
Beilstein/REAXYS Number:
4157942
EC Number:
MDL number:
Pricing and availability is not currently available.

assay

99% (titration)

form

solid

bp

315 °C (lit.)

mp

190-193 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Al](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Al/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

KILURZWTCGSYRE-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

UN 3467 6.1 / PGII

WGK Germany

3

Flash Point(F)

244.4 °F

Flash Point(C)

118 °C

Joanna Siuda et al.
Materials (Basel, Switzerland), 12(12) (2019-06-27)
It can be found that reaction mechanisms and interactions between wood and organosilicone compounds have not been sufficiently explored. The aim of the study was to determine bonds formed between either cellulose or lignin and methyltrimethoxysilane (MTMOS) during a catalytic...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.