Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0341

Sigma-Aldrich

HSP70 (EP377) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,050.00

$1,050.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$1,050.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$1,050.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

The heat shock proteins (HSPs) are a conserved family of ubiquitously expressed proteins that functions to protect cells from various environmental damage. Two proteins within the family, HSP70 and HSP27 demonstrate strong anti-apoptotic activity; its overexpression allow cells to survive in variable conditions. HSPs have been implicated in tumorigenesis – affecting cell cycle regulation, multidrug resistance and modulating p53 function. HSP70 has been studied in a variety of malignancies, including breast, lung, oral, prostate and uterine-cervical cancers. A majority of results associated HSP70 overexpression with poor prognosis and resistance to therapy. Recently, HSP70 diagnostic utility was established in identifying hepatocellular carcinoma (HCC). HSP70 was the most abundantly up-regulated gene in advanced HCC and its immunoreactivity was seen in the majority of HCC. Its sensitivity and specificity were determined as 74% and 98%, respectively. Furthermore, a three antibody panel with HSP70, Glypican 3, and Glutamine Synthetase is capable of distinguishing HCC from high grade dysplastic nodules with 100% specificity when 2 markers are positive.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &< 0.1% Sodium Azide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service