Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0351A

Sigma-Aldrich

Claudin-1 (EP420) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

Storage temp.

2-8°C

visualization

(membrane/cytoplasm)

General description

Claudin-1 (CLDN-1) is a member of the claudin integral membrane tight junction protein family. Claudins are responsible for regulating paracellular permeability, promoting cell adhesion and maintaining cell polarity and barrier function. Localized to the cytosol and membrane, Claudin-1 is broadly expressed in most tissues and is localized to most epithelial cells. Surrounding secretory epithelial cells of stroma components such as fibroblasts, pneumocytes and lymphocytes do not express Claudin-1. Claudin-1 expression is typically reduced in proliferating compared to quiescent epithelial cells. Claudin-1 was immuno-negative in 98% of prostate cancer cases and significantly decreased in colon carcinoma tissue compared to adjacent normal mucosa. Furthermore, Claudin-1 has demonstrated utility for identifying squamous cell carcinomas (SqCC). In lung tumors, Claudin-1 expression was detected in 78% of SqCC but only 17% of adenocarcinoma. Its upregulation is observed in pancreatic, thyroid, lung and oral SqCC.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &lt0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact [email protected]

For Technical Service please contact: 800-665-7284 or email: [email protected]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service