AMAB91337
Monoclonal Anti-HIST1H2BC antibody produced in mouse
Prestige Antibodies® Powered by Atlas Antibodies, clone CL5048, purified immunoglobulin, buffered aqueous glycerol solution
Synonym(s):
H2B, H2BFA, H2BFG, H2BFH, H2BFK, H2BFL
Select a Size
$598.00
Select a Size
About This Item
$598.00
conjugate
unconjugated
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
CL5048, monoclonal
product line
Prestige Antibodies® Powered by Atlas Antibodies
form
buffered aqueous glycerol solution
species reactivity
mouse, rat, human
technique(s)
immunohistochemistry: 1:20000-1:50000
western blot: 1 μg/mL
isotype
IgG2b
Ensembl | human accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... GJB6(10804)
Looking for similar products? Visit Product Comparison Guide
Immunogen
Sequence
VSEGTKAVTKYTSSK
Application
Features and Benefits
Every Prestige Antibody is tested in the following ways:
- IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
- Protein array of 364 human recombinant protein fragments.
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service