MilliporeSigma
  • APREST70355
All Photos(1)

APREST70355

Sigma-Aldrich

PrEST Antigen YY1

Prestige Antigens Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):
DELTA, INO80S, NF-E1, UCRBP, YIN-YANG-1

recombinant

expressed in E. coli

Quality Level

Assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 34 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

DLSDPKQLAEFARMKPRKIKEDDAPRTIACPHKGCTKMFRDNSAMRKHLHTHGPRVHVCAECGKAFVESSKLKRHQLVHTGEKPFQCTFEGCGKRFSLDFNLRTHVRIHTGDRPYVCPFDGCNKKFAQSTNLKSHILTH

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... YY1(7528)

Compare Similar Items

View Full Comparison

1 of 4

This Item
APREST90711APREST92124APREST81887
recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

recombinant

expressed in E. coli

assay

>80% (SDS-PAGE)

assay

>80% (SDS-PAGE)

assay

>80% (SDS-PAGE)

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

mol wt

predicted mol wt 34 kDa

mol wt

predicted mol wt 27 kDa

mol wt

predicted mol wt 28 kDa

mol wt

predicted mol wt 29 kDa

purified by

immobilized metal affinity chromatography (IMAC)

purified by

immobilized metal affinity chromatography (IMAC)

purified by

immobilized metal affinity chromatography (IMAC)

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

concentration

≥0.5 mg/mL

concentration

≥0.5 mg/mL

concentration

≥0.5 mg/mL

General description

Recombinant protein fragment of Human YY1 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

Application

Suitable as a blocking agent using corresponding antibodies.

Linkage

Corresponding Antibody HPA001119.

Physical form

Solution in 1 M urea-PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service