APREST89467
expressed in E. coli
>80% (SDS-PAGE)
buffered aqueous solution
predicted mol wt 33 kDa
immobilized metal affinity chromatography (IMAC)
≥0.5 mg/mL
QKEQEKAFFAQLQLDEETGEFLPIQPAQHIQSETSGSANYSQVAHIPKSDALYFDDCMQLLAQTFPFVDDNEVSSATFQSLVPDIPGHIESPVFIATNQAQSPETSVAQVAPVDLDGMQQDIEQVWEELLSIPELQCL
wet ice
−20°C
human ... NFE2L2(4780)
1 of 4
This Item | APREST88141 | APREST94189 | APREST92238 |
---|---|---|---|
recombinant expressed in E. coli | recombinant expressed in E. coli | recombinant expressed in E. coli | recombinant expressed in E. coli |
assay >80% (SDS-PAGE) | assay >80% (SDS-PAGE) | assay >80% (SDS-PAGE) | assay >80% (SDS-PAGE) |
form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous solution | form buffered aqueous solution |
mol wt predicted mol wt 33 kDa | mol wt predicted mol wt 26 kDa | mol wt predicted mol wt 22 kDa | mol wt predicted mol wt 30 kDa |
purified by immobilized metal affinity chromatography (IMAC) | purified by immobilized metal affinity chromatography (IMAC) | purified by immobilized metal affinity chromatography (IMAC) | purified by immobilized metal affinity chromatography (IMAC) |
concentration ≥0.5 mg/mL | concentration ≥0.5 mg/mL | concentration ≥0.5 mg/mL | concentration ≥0.5 mg/mL |
12 - Non Combustible Liquids
WGK 2
Not applicable
Not applicable
Enter Lot Number to search for Certificate of Analysis (COA).
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service