All Photos(1)

AV37859

Sigma-Aldrich

Anti-KLF2 antibody produced in rabbit

IgG fraction of antiserum

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

39 kDa

species reactivity

rat, pig, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... KLF2(16598)

General description

Krüppel-like factor 2 (KLF2) is a multifunction transcriptional regulator with growth-inhibitory, pro-apoptotic and anti-angiogenic activity towards monocytic cells. KLF2 modulates gene expression in endothelial cells. It is a positive regulator of embryonic β-globin gene expression.

Specificity

Anti-KLF2 polyclonal antibody reacts with human, mouse, rat, and pig Krüppel-like factor 2 proteins.

Immunogen

The immunogen for anti-KLF2 antibody: synthetic peptide derected towards the middle region of human KLF2

Application

Anti-KLF2 polyclonal antibody is used to tag Krüppel-like factor 2 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of Krüppel-like factor 2 in gene expression and cell processes such as growth, differentiation and apoptosis.

Sequence

Synthetic peptide located within the following region: IPASGPRNPFPPPFGPGPSFGGPGPALHYGPPAPGAFGLFEDAAAAAAAL

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service