MilliporeSigma
All Photos(1)

B1900

Sigma-Aldrich

Boc-Glu(OcHx)-OH

Synonym(s):
Boc-L-glutamic acid γ-cyclohexyl ester, Boc-L-glutamic acid 5-cyclohexyl ester
Linear Formula:
C6H11OCOCH2CH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
329.39
Beilstein:
3595818
MDL number:

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCC(=O)OC1CCCCC1)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service