Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B7133

Sigma-Aldrich

Basic Yellow 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClN2O2
CAS Number:
Molecular Weight:
372.89
Colour Index Number:
48055
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ~20%

SMILES string

[Cl-].COc1ccc(NC=CC2=[N+](C)c3ccccc3C2(C)C)c(OC)c1

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service